Thermo Scientific Chemicals

4-(Trifluoromethyl)benzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: 208920500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(Trifluoromethyl)benzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: 208920500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 208920500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
455-19-6
IUPAC Name
4-(trifluoromethyl)benzaldehyde
Molecular Formula
C8H5F3O
InChI Key
BEOBZEOPTQQELP-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=CC=C(C=O)C=C1
GC
>=97.5 %
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Refractive index
1.4625 to 1.4645 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text