Thermo Scientific Chemicals

1-Propanephosphonic acid cyclic anhydride, 50 wt.% sol. in ethyl acetate, Thermo Scientific Chemicals

Catalog number: 209010500
Each
Thermo Scientific Chemicals

1-Propanephosphonic acid cyclic anhydride, 50 wt.% sol. in ethyl acetate, Thermo Scientific Chemicals

Catalog number: 209010500
Each
Catalog number: 209010500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
68957-94-8
IUPAC Name
tripropyl-1,3,5,2λ⁵,4λ⁵,6λ⁵-trioxatriphosphinane-2,4,6-trione
Molecular Formula
C9H21O6P3
InChI Key
PAQZWJGSJMLPMG-UHFFFAOYSA-N
SMILES
CCCP1(=O)OP(=O)(CCC)OP(=O)(CCC)O1
Appearance (Color)
Clear colorless to amber
Assay
>=50 % w/w Solids
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text