Cyclobutylamine, 98%, Thermo Scientific Chemicals
Cyclobutylamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclobutylamine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mL
5 mL
Packaging:
Glass bottle
Catalog number 209110010
Price (USD)/ Each
57.90
-
Add to cart
Quantity:
1 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
57.90
Add to cart
Cyclobutylamine, 98%, Thermo Scientific Chemicals
Catalog number209110010
Price (USD)/ Each
57.90
-
Add to cart
Chemical Identifiers
CAS2516-34-9
IUPAC Namecyclobutanamine
Molecular FormulaC4H9N
InChI KeyKZZKOVLJUKWSKX-UHFFFAOYSA-N
SMILESNC1CCC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4360 to 1.4400 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only