Thermo Scientific Chemicals

3-Nitrobenzenesulfonic acid, sodium salt, 99%, Thermo Scientific Chemicals

Catalog number: 209120010
1 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

3-Nitrobenzenesulfonic acid, sodium salt, 99%, Thermo Scientific Chemicals

Catalog number: 209120010
1 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 209120010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
127-68-4
IUPAC Name
sodium 3-nitrobenzene-1-sulfonate
Molecular Formula
C6H4NNaO5S
InChI Key
LJRGBERXYNQPJI-UHFFFAOYSA-M
SMILES
[Na+].[O-][N+](=O)C1=CC=CC(=C1)S([O-])(=O)=O
Appearance (Form)
Powder or crystalline powder
Appearance (Color)
White to yellow
HPLC
>=96.0 %
Infrared spectrum
Conforms
Water
=<3.0 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text