Chemical Identifiers
CAS17685-52-8
IUPAC Namedodecakis(methanidylidyneoxidanium) triiron
Molecular FormulaC12Fe3O12
InChI KeyHLYRMDDXFDINCB-UHFFFAOYSA-N
SMILES[Fe].[Fe].[Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
View more