Thermo Scientific Chemicals

Cyclopentadienyliron dicarbonyl dimer, 99%, Thermo Scientific Chemicals

Catalog number: 209300250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Cyclopentadienyliron dicarbonyl dimer, 99%, Thermo Scientific Chemicals

Catalog number: 209300250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 209300250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12154-95-9
IUPAC Name
bis([(cyclopenta-2,4-dien-1-yl)ferrio]methanone); bis(methanidylidyneoxidanium)
Molecular Formula
C14H10Fe2O4
InChI Key
XJSJEKXJJGHIGW-UHFFFAOYSA-N
SMILES
[C-]#[O+].[C-]#[O+].O=[C-][Fe+]C1C=CC=C1.O=[C-][Fe+]C1C=CC=C1
Elemental Analysis
46.8 to 48.2 % (C)
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Appearance (Color)
Red-purple to dark brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text