Chemical Identifiers
CAS12154-95-9
IUPAC Namebis([(cyclopenta-2,4-dien-1-yl)ferrio]methanone); bis(methanidylidyneoxidanium)
Molecular FormulaC14H10Fe2O4
InChI KeyXJSJEKXJJGHIGW-UHFFFAOYSA-N
SMILES[C-]#[O+].[C-]#[O+].O=[C-][Fe+]C1C=CC=C1.O=[C-][Fe+]C1C=CC=C1
View more