1,3,5-Benzenetricarboxylic acid triallyl ester, 98%, Thermo Scientific Chemicals
1,3,5-Benzenetricarboxylic acid triallyl ester, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3,5-Benzenetricarboxylic acid triallyl ester, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 209350010
Price (USD)/ Each
40.11
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
40.11
Add to cart
1,3,5-Benzenetricarboxylic acid triallyl ester, 98%, Thermo Scientific Chemicals
Catalog number209350010
Price (USD)/ Each
40.11
-
Add to cart
Chemical Identifiers
IUPAC Name1,3,5-tris(prop-2-en-1-yl) benzene-1,3,5-tricarboxylate
Molecular FormulaC18H18O6
InChI KeyVOSUIKFOFHZNED-UHFFFAOYSA-N
SMILESC=CCOC(=O)C1=CC(=CC(=C1)C(=O)OCC=C)C(=O)OCC=C
Molecular Weight (g/mol)330.34
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow
Infrared spectrumConforms
Appearance (Form)Liquid or low melting solid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only