Thermo Scientific Chemicals

Tetrapropylammonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: 209375000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Tetrapropylammonium bromide, 98%, Thermo Scientific Chemicals

Catalog number: 209375000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 209375000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1941-30-6
IUPAC Name
tetrapropylazanium bromide
Molecular Formula
C12H28BrN
InChI Key
BGQMOFGZRJUORO-UHFFFAOYSA-M
SMILES
[Br-].CCC[N+](CCC)(CCC)CCC
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals
Titration Argentometric
97.5 to 102.5 %
Appearance (Color)
White to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text