Thermo Scientific Chemicals

Trimethylsilyl trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: 209442500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

Trimethylsilyl trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals

Catalog number: 209442500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 209442500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27607-77-8
IUPAC Name
trimethylsilyl trifluoromethanesulfonate
Molecular Formula
C4H9F3O3SSi
InChI Key
FTVLMFQEYACZNP-UHFFFAOYSA-N
SMILES
C[Si](C)(C)OS(=O)(=O)C(F)(F)F
Appearance (Color)
Clear colorless to light brown
Titration with NaOH
>=98.5 %
Infrared spectrum
Conforms
Refractive index
1.3600 to 1.3620 (20°C, 589 nm)
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text