Bis(acetonitrile)palladium(II) chloride, 99%, Thermo Scientific Chemicals
Bis(acetonitrile)palladium(II) chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(acetonitrile)palladium(II) chloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
5 g
Packaging:
Glass bottle
Catalog number 209455000
Price (USD)/ Each
-
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14592-56-4
IUPAC Namebis(acetonitrile); dichloropalladiumbis(ylium)
Molecular FormulaC4H6Cl2N2Pd
InChI KeyXGPJSRXOBCGBMN-UHFFFAOYSA-L
SMILESCC#N.CC#N.Cl[Pd++]Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to rust brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Elemental Analysis>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only