Silver sulfate, 99%, extra pure, Thermo Scientific Chemicals
Silver sulfate, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver sulfate, 99%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 209470100
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
Add to cart
Silver sulfate, 99%, extra pure, Thermo Scientific Chemicals
Catalog number209470100
Price (USD)/ Each
105.65
Online exclusive
117.00 
Save 11.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10294-26-5
IUPAC Namedisilver(1+) sulfuric acid
Molecular FormulaAg2H2O4S
InChI KeyYPNVIBVEFVRZPJ-UHFFFAOYSA-N
SMILES[Ag+].[Ag+].OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=98.5 %
Appearance (Color)White to off-white
Insoluble matter=<0.02 % (in water and AgCl)
Iron (Fe)=<10 ppm
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only