Thermo Scientific Chemicals

4-(Diethylamino)salicylaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: 209480250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(Diethylamino)salicylaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: 209480250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 209480250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
17754-90-4
IUPAC Name
4-(diethylamino)-2-hydroxybenzaldehyde
Molecular Formula
C11H15NO2
InChI Key
XFVZSRRZZNLWBW-UHFFFAOYSA-N
SMILES
CCN(CC)C1=CC=C(C=O)C(O)=C1
Appearance (Color)
Pink to light brown to purple
GC
>=97.5 %
Infrared spectrum
Conforms
Appearance (Form)
Powder
Melting point
60°C to 64°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text