Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number 209531000
Price (USD)/ Each
56.65
Online exclusive
62.50 
Save 5.85 (9%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)/ Each
56.65
Online exclusive
62.50 
Save 5.85 (9%)
Add to cart
Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
Catalog number209531000
Price (USD)/ Each
56.65
Online exclusive
62.50 
Save 5.85 (9%)
-
Add to cart
Chemical Identifiers
CAS1730-25-2
IUPAC Namebromo(prop-2-en-1-yl)magnesium
Molecular FormulaC3H5BrMg
InChI KeyFEMBXICCJNZMMC-UHFFFAOYSA-M
SMILESBr[Mg]CC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Yellow-green to grey
Titration Grignard0.9 to 1.1 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only