Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | 2095 Phosphate 50 ppm Standard, 5L |
Type | Phosphate Standard |
Unit Size | Each |
Description | 2095 Phosphate 2 ppm Standard, 5L |
Type | Standards |
Unit Size | Each |
Description | 2095 Phosphate 100 ppm Standard, 5L |
Type | Phosphate Standard |
Unit Size | Each |
Description | 2095 Phosphate Reagent, Molybdenum Blue 0-10ppm |
Type | Reagent |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
209550 | Each | 2095 Phosphate 50 ppm Standard, 5L | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
209502 | Each | 2095 Phosphate 2 ppm Standard, 5L | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
209510 | Each | 2095 Phosphate 100 ppm Standard, 5L | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
2095REB | Each | 2095 Phosphate Reagent, Molybdenum Blue 0-10ppm | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|