Lithium bis(trimethylsilyl)amide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Lithium bis(trimethylsilyl)amide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium bis(trimethylsilyl)amide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Catalog number 209551000
Price (USD)/ Each
52.65
Online exclusive
62.00 
Save 9.35 (15%)
-
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)/ Each
52.65
Online exclusive
62.00 
Save 9.35 (15%)
Add to cart
Lithium bis(trimethylsilyl)amide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Catalog number209551000
Price (USD)/ Each
52.65
Online exclusive
62.00 
Save 9.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namelithium(1+) bis(trimethylsilyl)azanide
Molecular FormulaC6H18LiNSi2
InChI KeyYNESATAKKCNGOF-UHFFFAOYSA-N
SMILES[Li+].C[Si](C)(C)[N-][Si](C)(C)C
Molecular Weight (g/mol)167.33
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to amber
Strength0.90 to 1.10 M
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only