Thermo Scientific Chemicals

Lithium bis(trimethylsilyl)amide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 209558000
800 mL, Each
Thermo Scientific Chemicals

Lithium bis(trimethylsilyl)amide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 209558000
800 mL, Each
Quantity
Catalog number: 209558000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4039-32-1, 109-99-9
Appearance (Color)
Light yellow to amber
Strength
0.90 to 1.10 M
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text