Benzaldehyde dimethyl acetal, 98%, Thermo Scientific Chemicals
Benzaldehyde dimethyl acetal, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Benzaldehyde dimethyl acetal, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 209791000
Price (USD)/ Each
40.65
Online exclusive
47.10 
Save 6.45 (14%)
-
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
40.65
Online exclusive
47.10 
Save 6.45 (14%)
Add to cart
Benzaldehyde dimethyl acetal, 98%, Thermo Scientific Chemicals
Catalog number209791000
Price (USD)/ Each
40.65
Online exclusive
47.10 
Save 6.45 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Name(dimethoxymethyl)benzene
Molecular FormulaC9H12O2
InChI KeyHEVMDQBCAHEHDY-UHFFFAOYSA-N
SMILESCOC(OC)C1=CC=CC=C1
Molecular Weight (g/mol)152.19
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Clear colorless
Refractive index1.4910 to 1.4940 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only