Chemical Identifiers
CAS56602-33-6
IUPAC Name(1H-1,2,3-benzotriazol-1-yloxy)tris(dimethylamino)phosphanium; hexafluoro-λ⁵-phosphanuide
Molecular FormulaC12H22F6N6OP2
InChI KeyMGEVGECQZUIPSV-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.CN(C)[P+](ON1N=NC2=CC=CC=C12)(N(C)C)N(C)C
View more