Thermo Scientific Chemicals

tert-Butyldimethylsilyl trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 209860050
5 mL, Glass bottle, Each
Thermo Scientific Chemicals

tert-Butyldimethylsilyl trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals

Catalog number: 209860050
5 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 209860050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
69739-34-0
IUPAC Name
tert-butyldimethylsilyl trifluoromethanesulfonate
Molecular Formula
C7H15F3O3SSi
InChI Key
WLLIXJBWWFGEHT-UHFFFAOYSA-N
SMILES
CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F
Infrared spectrum
Conforms
Refractive index
1.3850 to 1.3880 (20°C, 589 nm)
Appearance (Form)
Liquid
Assay
>=97.5 %
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text