tert-Butyl N-hydroxycarbamate, 98+%, Thermo Scientific Chemicals
tert-Butyl N-hydroxycarbamate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butyl N-hydroxycarbamate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 209870050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS36016-38-3
IUPAC Nametert-butyl N-hydroxycarbamate
Molecular FormulaC5H11NO3
InChI KeyDRDVJQOGFWAVLH-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point53°C to 57°C
Infrared spectrumConforms
Appearance (Color)White to light pink
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only