Thermo Scientific Chemicals

(-)-Camphanic acid chloride, 98%, Thermo Scientific Chemicals

Catalog number: 209890050
5 g, Plastic bottle, Each
Thermo Scientific Chemicals

(-)-Camphanic acid chloride, 98%, Thermo Scientific Chemicals

Catalog number: 209890050
5 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 209890050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
39637-74-6
IUPAC Name
(1S,4R)-4,7,7-trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride
Molecular Formula
C10H13ClO3
InChI Key
PAXWODJTHKJQDZ-VHSXEESVSA-N
SMILES
CC1(C)[C@@]2(C)CC[C@@]1(OC2=O)C(Cl)=O
Appearance (Color)
White to beige to grey
Infrared spectrum
Conforms
Titration RCOCl
>=97.5 %
Appearance (Form)
Powder or crystals or flakes
Melting point
67°C to 71°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text