Thermo Scientific Chemicals

N,N-Dimethyltrimethylsilylamine, 97%, Thermo Scientific Chemicals

Catalog number: 209970100
10 mL, Glass bottle, Each
Thermo Scientific Chemicals

N,N-Dimethyltrimethylsilylamine, 97%, Thermo Scientific Chemicals

Catalog number: 209970100
10 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 209970100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2083-91-2
IUPAC Name
dimethyl(trimethylsilyl)amine
Molecular Formula
C5H15NSi
InChI Key
KAHVZNKZQFSBFW-UHFFFAOYSA-N
SMILES
CN(C)[Si](C)(C)C
Appearance (Color)
Clear colorless to light yellow
Titration with HClO4
>=96.0 %
Appearance (Form)
Liquid
Refractive index
1.3960 to 1.3980 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text