Pierce™ Protein A IgG Binding Buffer
Pierce™ Protein A IgG Binding Buffer
Thermo Scientific™

Pierce™ Protein A IgG Binding Buffer

Thermo Scientific Pierce IgG Binding and Elution Buffers ensure high-yield, nondenaturing antibody purification with IgG affinity purification supports, such asRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
210011 L
210073.75 L
Catalog number 21001
Price (USD)
122.65
Online Exclusive
135.00
Save 12.35 (9%)
Each
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)
122.65
Online Exclusive
135.00
Save 12.35 (9%)
Each
Add to cart
Thermo Scientific Pierce IgG Binding and Elution Buffers ensure high-yield, nondenaturing antibody purification with IgG affinity purification supports, such as Protein A and Protein G agarose beads. The Protein A IgG Binding Buffer is a phosphate-based buffer containing glycerol and EDTA, formulated for antibody purification using Protein A.

Shared features of all Thermo Scientific Pierce IgG Binding and Elution Buffers:

Ready to use—carefully formulated and prefiltered 1X solutions
Three binding buffers—optimized for Protein A, Protein G or Protein A/G
Two elution buffer options—acidic (pH 2.8) or high salt (near-neutral)
Versatile—elution buffers are appropriate for antibody-antigen affinity purification

Products available for each antibody purification method:
Protein A IgG Binding Buffer: pH 8.0; phosphate-based; contains glycerol and EDTA
Protein A/G IgG Binding Buffer: pH 8.0; phosphate-based; contains EDTA
Protein G IgG Binding Buffer: pH 5.0; contains 0.02% sodium azide
IgG Elution Buffer: pH 2.8; amine-based
Gentle Ag/Ab Binding Buffer: pH 8.0; phosphate-free; contains 0.05% sodium azide
Gentle Ag/Ab Elution Buffer: pH 6.6; high salt (ionic strength)

Related Products
Pierce™ IgG Elution Buffer
Pierce™ Protein G IgG Binding Buffer
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid
Quantity1 L
Reagent TypeBinding Buffer
Product LinePierce™
Product TypeBinding Buffer
pH8
Unit SizeEach
Contents & Storage
4°C
Learn about immunopeptidomics basics
Whether you're new to or experienced with proteomics, we're here to help.


Learn about immunopeptidomics basics ›
Follow 5-step sample prep workflow ›

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link