Thermo Scientific Chemicals

2-(Methylthio)ethanol, 99%, Thermo Scientific Chemicals

Catalog number: 210100250
25 mL, Glass bottle, Each
Thermo Scientific Chemicals

2-(Methylthio)ethanol, 99%, Thermo Scientific Chemicals

Catalog number: 210100250
25 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 210100250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5271-38-5
IUPAC Name
2-(methylsulfanyl)ethan-1-ol
Molecular Formula
C3H8OS
InChI Key
WBBPRCNXBQTYLF-UHFFFAOYSA-N
SMILES
CSCCO
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.4934 to 1.4954 (20°C, 589 nm)
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text