Thermo Scientific Chemicals

Silver perchlorate hydrate, 99%, Thermo Scientific Chemicals

Catalog number: 210211000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Silver perchlorate hydrate, 99%, Thermo Scientific Chemicals

Catalog number: 210211000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 210211000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14242-05-8
IUPAC Name
silver(1+) perchlorate
Molecular Formula
AgClO4
InChI Key
YDHABVNRCBNRNZ-UHFFFAOYSA-M
SMILES
[Ag+].[O-][Cl](=O)(=O)=O
Appearance (Form)
Crystalline powder or crystals
Sulfate (SO4)
=<10 ppm
Appearance (Color)
White
Water
=<10 % (K.F.)
Chloride (Cl)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text