Orion™ 2100 Series pH ORP and Conductivity Analyzer
Catalog number: 2102PH2
Have Questions?
Thermo Scientific™
Orion™ 2100 Series pH ORP and Conductivity Analyzer
Catalog number: 2102PH2
Have Questions?
Measure pH/ORP and conductivity accurately and reliably in the harshest environments with the Thermo Scientific™ Orion™ 2100 Series pH ORP and Conductivity Analyzer. Designed for the highest accuracy and performance, this analyzer has flexible measurement parameters for single, dual or combined pH ORP. Conductivity inputs are offered with optional digital communications.
2 x 110250 ORP electrodes and ORP standard (967901)
Keypad Type
Membrane
Max. Relative Humidity
5 to 95%, non-condensing
Mounting Options
Pipe, Panel or Wall Mount
Options for Channel 1
pH/ORP
Options for Channel 2
pH/ORP
Range (mV)
±1999mV
Range (Resistivity)
2ohm to 100Mohm
Resolution
0.1; 0.01
Resolution (mV)
1mV
Resolution (Resistivity)
2ohms/cm
Resolution (Temperature)
0.1°C
Temperature (English) Operating
41°F to 113°F
USB Interface
No
Length (Metric)
186 mm
Width (Metric)
144 mm
Height (Metric)
144 mm
Voltage
85 to 132 VAC; 170 to 264 VAC
Amperage
5A
Frequency
0 to 60 Hz
Unit Size
Each
Showing 10 of 15Show more
Features:
Single channel or dual channel configurations
Optional digital communication boards
Provides cation and ammonia/ETA compensation for customizing to your plant's requirement
2100 Series pH Analyzer Consumables Ordering Information:
pH Consumables Kit — pH 4.01, 7.00 and 10.01 Buffers and Process Thermo Scientific™ Ross™ Electrode Filling Solution, Cat. No. 2001CP
pH 4.01 Buffer, 475mL, Cat. No. 910104
pH 7.00 Buffer, 475mL, Cat. No. 910107
pH 10.01 Buffer, 475mL, Cat. No. 910110
Process Ross Electrode Filling Solution, 5 x 60mL, Cat. No. 2001FS
Process Ross Electrode Filling Solution, 4 x 120mL, Cat. No. 2001F4
pH Electrode Cleaning Solution A for Protein Removal, 4 x 30mL, Cat. No. 900021
pH Electrode Cleaning Solution B for Bacteria Removal, 4 x 60mL, Cat. No. 900022
pH Electrode Cleaning Solution C for General Cleaning, 4 x 30mL, Cat. No. 900023
pH Electrode Cleaning Solution D for Oil and Grease Removal, 4 x 60mL, Cat. No. 900024
2100 Series Conductivity Analyzer Consumables Ordering Information:
111.9mS/cm Conductivity Standard, 5 x 60mL, Cat. No. 011005
12.9mS/cm Conductivity Standard, 5 x 60mL, Cat. No. 011006
1413μS/cm Conductivity Standard, 5 x 60mL, Cat. No. 011007
100μS/cm Conductivity Standard, 5 x 60mL, Cat. No. 011008
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.