Thermo Scientific Chemicals

Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: 210490250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Methoxylamine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: 210490250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 210490250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
593-56-6
IUPAC Name
hydrogen O-methylhydroxylamine chloride
Molecular Formula
CH6ClNO
InChI Key
XNXVOSBNFZWHBV-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CON
Appearance (Color)
White to light yellow
Melting point
148°C to 153°C
Appearance (Form)
Crystalline powder
Titration with HClO4
98.0 to 102.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text