Cyclohexane, extra pure, Thermo Scientific Chemicals
Cyclohexane, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclohexane, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
10 L
Packaging:
View all
Glass Bottle
Catalog number 210570025
Price (USD)/ Each
123.65
Online exclusive
137.00 
Save 13.35 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
123.65
Online exclusive
137.00 
Save 13.35 (10%)
Add to cart
Cyclohexane, extra pure, Thermo Scientific Chemicals
Catalog number210570025
Price (USD)/ Each
123.65
Online exclusive
137.00 
Save 13.35 (10%)
-
Add to cart
Chemical Identifiers
CAS110-82-7
IUPAC Namecyclohexane
Molecular FormulaC6H12
InChI KeyXDTMQSROBMDMFD-UHFFFAOYSA-N
SMILESC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
Residue after evaporation=<0.005 %
Water=<0.01 %
Color scale=<10 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only