Thermo Scientific Chemicals

Hexanes, technical, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 210590050
5 L, Plastic Drum, Each
Thermo Scientific Chemicals

Hexanes, technical, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 210590050
5 L, Plastic Drum, Each
Quantity
Packaging
Catalog number: 210590050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
92112-69-1, 110-54-3
Color scale
=<10 APHA
Appearance (Form)
Clear liquid
Residue after evaporation
=<0.002 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text