Thermo Scientific Chemicals

Deuterium chloride, for NMR, 1M solution in D2O, 99.8 atom% D, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 210601000
100 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Deuterium chloride, for NMR, 1M solution in D2O, 99.8 atom% D, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 210601000
100 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 210601000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7698-05-7
IUPAC Name
proton chloride
Molecular Formula
ClH
InChI Key
VEXZGXHMUGYJMC-FTGQXOHASA-N
SMILES
[1H+].[Cl-]
Appearance (Color)
Colorless
Appearance (Form)
Liquid
D-Enrichment
>=99.75 %
Strength
0.95 to 1.05 M

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text