Silver nitrite, 99%, Thermo Scientific Chemicals
Silver nitrite, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver nitrite, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 210690100
Price (USD)/ Each
64.65
Online exclusive
71.20 
Save 6.55 (9%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
64.65
Online exclusive
71.20 
Save 6.55 (9%)
Add to cart
Silver nitrite, 99%, Thermo Scientific Chemicals
Catalog number210690100
Price (USD)/ Each
64.65
Online exclusive
71.20 
Save 6.55 (9%)
-
Add to cart
Chemical Identifiers
CAS7783-99-5
IUPAC Namesilver(1+) nitrite
Molecular FormulaAgNO2
InChI KeyKKKDGYXNGYJJRX-UHFFFAOYSA-M
SMILES[Ag+].[O-]N=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green
Appearance (Form)Powder or crystals and/or chunks
Titration Argentometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only