2-Mesitylmagnesium bromide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
2-Mesitylmagnesium bromide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Mesitylmagnesium bromide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 210731000
Price (USD)/ Each
110.65
Online exclusive
123.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
110.65
Online exclusive
123.00 
Save 12.35 (10%)
Add to cart
2-Mesitylmagnesium bromide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Catalog number210731000
Price (USD)/ Each
110.65
Online exclusive
123.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2633-66-1
IUPAC Namebromo(3,4,5-trimethylphenyl)magnesium
Molecular FormulaC9H11BrMg
InChI KeyVZMGGVXGYWKQPB-UHFFFAOYSA-M
SMILESCC1=CC([Mg]Br)=CC(C)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to brown
Appearance (Form)Solution
Titration Grignard0.95 to 1.10 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only