Methyl 2,2-dimethyl-3-hydroxypropionate, 98+%, Thermo Scientific Chemicals
Methyl 2,2-dimethyl-3-hydroxypropionate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 2,2-dimethyl-3-hydroxypropionate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 210910250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namemethyl 3-hydroxy-2,2-dimethylpropanoate
Molecular FormulaC6H12O3
InChI KeyKJRFTNVYOAGTHK-UHFFFAOYSA-N
SMILESCOC(=O)C(C)(C)CO
Molecular Weight (g/mol)132.16
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4280 to 1.4310 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only