Thermo Scientific Chemicals

Calcium acetate hydrate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 211050010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Calcium acetate hydrate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 211050010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 211050010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
114460-21-8
IUPAC Name
calcium diacetate
Molecular Formula
C4H6CaO4
InChI Key
VSGNNIFQASZAOI-UHFFFAOYSA-L
SMILES
[Ca++].CC([O-])=O.CC([O-])=O
Appearance (Color)
White
Infrared spectrum
Conforms
Water
=<7.0 % (K.F.)
Arsenic (As)
=<2 ppm
Heavy metals (as Pb)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text