Chemical Identifiers
CAS299-28-5
IUPAC Namecalcium bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
Molecular FormulaC12H22CaO14
InChI KeyNEEHYRZPVYRGPP-IYEMJOQQSA-L
SMILES[Ca++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
View more