Magnesium carbonate basic, light, for analysis, 40.0-45.0% Magnesium oxide, Thermo Scientific Chemicals
Magnesium carbonate basic, light, for analysis, 40.0-45.0% Magnesium oxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium carbonate basic, light, for analysis, 40.0-45.0% Magnesium oxide, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
1 kg
Packaging:
Plastic drum
Catalog number 211070025
Price (USD)/ Each
-
Quantity:
2.5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS39409-82-0
IUPAC Namepentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
Molecular FormulaC4H10Mg5O18
InChI KeyYMKSTRLFWJQKFP-UHFFFAOYSA-D
SMILESO.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Soluble matter=<1 % (in water)
Insoluble matter=<0.05 % (in acetic acid)
Heavy metals (as Pb)=<20 ppm
Chloride (Cl)=<700 ppm
Sulfate (SO4)=<3000 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only