Nickel(II) sulfate hexahydrate, 99%, for analysis, Thermo Scientific Chemicals
Nickel(II) sulfate hexahydrate, 99%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel(II) sulfate hexahydrate, 99%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
10 kg
Packaging:
View all
Plastic bottle
Catalog number 211080025
Price (USD)/ Each
476.65
Online exclusive
560.00 
Save 83.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
476.65
Online exclusive
560.00 
Save 83.35 (15%)
Add to cart
Nickel(II) sulfate hexahydrate, 99%, for analysis, Thermo Scientific Chemicals
Catalog number211080025
Price (USD)/ Each
476.65
Online exclusive
560.00 
Save 83.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namenickel(2+) hexahydrate sulfate
Molecular FormulaH12NiO10S
InChI KeyRRIWRJBSCGCBID-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O
Molecular Weight (g/mol)262.84
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue-green
Appearance (Form)Crystals
Titration Complexometric98.5 to 101.5 %
pH4 to 6 (5 % at 20°C)
Chloride (Cl)=<50 ppm
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only