Thermo Scientific Chemicals

Nickel(II) sulfate hexahydrate, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 211081000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Nickel(II) sulfate hexahydrate, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 211081000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 211081000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10101-97-0
IUPAC Name
nickel(2+) hexahydrate sulfate
Molecular Formula
H12NiO10S
InChI Key
RRIWRJBSCGCBID-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Ni++].[O-]S([O-])(=O)=O
Appearance (Color)
Blue-green
Appearance (Form)
Crystals
Titration Complexometric
98.5 to 101.5 %
pH
4 to 6 (5 % at 20°C)
Chloride (Cl)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text