Potassium ferrocyanide trihydrate, 99+%, for analysis
Potassium ferrocyanide trihydrate, 99+%, for analysis
Potassium ferrocyanide trihydrate, 99+%, for analysis
Potassium ferrocyanide trihydrate, 99+%, for analysis
Thermo Scientific Chemicals

Potassium ferrocyanide trihydrate, 99+%, for analysis

CAS: 14459-95-1 | C6H6FeK4N6O3 | 422.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 211095000
Price (USD)
80.65
Special offer
Online exclusive
Ends: 26-Sep-2025
99.80
Save 19.15 (19%)
Each
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
80.65
Special offer
Online exclusive
Ends: 26-Sep-2025
99.80
Save 19.15 (19%)
Each
Add to cart
Chemical Identifiers
CAS14459-95-1
IUPAC Nametetrapotassium hexacyanoironbis(ylium) trihydrate
Molecular FormulaC6H6FeK4N6O3
InChI KeyNLYKFUFIUWPPIZ-UHFFFAOYSA-N
SMILESO.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Titration with KMnO4>=99.0 %
Sulfate (SO4)=<100 ppm
Chloride (Cl)=<100 ppm
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Share catalog number, name or link