Molybdenyl acetylacetonate, 97%, Thermo Scientific Chemicals
Molybdenyl acetylacetonate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdenyl acetylacetonate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Packaging:
Glass bottle
Catalog number 211142500
Price (USD)/ Each
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS17524-05-9
IUPAC Namedioxomolybdenumbis(ylium) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14MoO6
InChI KeySKMUJBBRXZPAJY-VGKOASNMSA-L
SMILESO=[Mo++]=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to grey-green to light brown
Infrared spectrumConforms
Elemental Analysis>=96.0 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only