Thermo Scientific Chemicals

N-(Benzyloxycarbonyloxy)succinimide, 98%, Thermo Scientific Chemicals

Catalog number: 211171000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

N-(Benzyloxycarbonyloxy)succinimide, 98%, Thermo Scientific Chemicals

Catalog number: 211171000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 211171000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13139-17-8
IUPAC Name
benzyl 2,5-dioxopyrrolidin-1-yl carbonate
Molecular Formula
C12H11NO5
InChI Key
MJSHDCCLFGOEIK-UHFFFAOYSA-N
SMILES
O=C(OCC1=CC=CC=C1)ON1C(=O)CCC1=O
Infrared spectrum
Conforms
Appearance (Color)
White to off-white
HPLC
>=97.5 %
Appearance (Form)
Crystalline powder
Melting point
77°C to 82°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text