Thermo Scientific Chemicals

(-)-Diisopropyl D-tartrate, 99%, Thermo Scientific Chemicals

Catalog number: 211180500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

(-)-Diisopropyl D-tartrate, 99%, Thermo Scientific Chemicals

Catalog number: 211180500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 211180500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
62961-64-2
IUPAC Name
1,4-bis(propan-2-yl) (2R,3S)-2,3-dihydroxybutanedioate
Molecular Formula
C10H18O6
InChI Key
XEBCWEDRGPSHQH-OCAPTIKFSA-N
SMILES
CC(C)OC(=O)[C@@H](O)[C@@H](O)C(=O)OC(C)C
Appearance (Color)
Clear colorless
GC
>=98.5 %
Specific optical rotation
-16° to -18.5° (20°C, 589 nm) (neat)
Infrared spectrum
Conforms
Refractive index
1.4380 to 1.4400 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text