Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | DIPA Reagent - for 60 days operation, includes 1 x 0.8L DIPA reagent and diffusion tubing kit (Cat. No. 211194) |
Type | Tubing |
Unit Size | Each |
Description | Acetic Acid Reagent for 1810 Ammonia Analyzers, 1L |
Type | Reagent |
Unit Size | Each |
Description | Ammonia Reagent for 1811 Sodium Analyzers, 2L |
Type | Reagent |
Unit Size | Each |
Description | Iodide Reagent for 1818 Oxygen Scavenger Analyzers, 3 x 1L with 3 Diffusion Tubings |
Type | Reagent |
For Use With (Equipment) | Thermo Scientific™ 1800 Series Analyzer |
Unit Size | Each |
Description | Formic Acid Reagent for 2109XP Fluoride, 2117XP Chloride and 2120XP Calcium Hardness Analyzers, 1L with Diffusion Tubing |
Type | Reagent |
Unit Size | Each |
Description | DIPA Reagent Pack - for one year operation, includes 6 x 0.8L DIPA reagent and 6 diffusion tubing kits (6 x 211194) |
Type | Tubing |
For Use With (Equipment) | Orion™ Online Monitors |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
211190 | Each | DIPA Reagent - for 60 days operation, includes 1 x 0.8L DIPA reagent and diffusion tubing kit (Cat. No. 211194) | Request A Quote | - | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
181030 | Each | Acetic Acid Reagent for 1810 Ammonia Analyzers, 1L | 257.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
181130 | Each | Ammonia Reagent for 1811 Sodium Analyzers, 2L | 214.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
181811 | Each | Iodide Reagent for 1818 Oxygen Scavenger Analyzers, 3 x 1L with 3 Diffusion Tubings | Request A Quote | - | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
182011 | Each | Formic Acid Reagent for 2109XP Fluoride, 2117XP Chloride and 2120XP Calcium Hardness Analyzers, 1L with Diffusion Tubing | 228.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
211191 | Each | DIPA Reagent Pack - for one year operation, includes 6 x 0.8L DIPA reagent and 6 diffusion tubing kits (6 x 211194) | Request A Quote | - | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Reagents for the Orion 2109XP, 2110XP, 2111LL, 2111XP, 2117XP, 2118XP, 2120XP, 1809, 1810, 1811AO, 1811EL, 1817HL, 1817LL, 1818 and 1820 analyzers.
Warranty: Out of box