Thermo Scientific Chemicals

Xylenol Orange, sodium salt

Catalog number: 211220050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Xylenol Orange, sodium salt

Catalog number: 211220050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 211220050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3618-43-7
IUPAC Name
tetrasodium 2-[({5-[3-(3-{[bis(carboxylatomethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-2,2-dioxo-3H-1,2λ⁶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxylatomethyl)amino]acetate
Molecular Formula
C31H28N2Na4O13S
InChI Key
FXYMNKLSOQONJB-UHFFFAOYSA-J
SMILES
[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC(CN(CC([O-])=O)CC([O-])=O)=C1O)C1(C2=CC=CC=C2OS1(=O)=O)C1=CC(C)=C(O)C(CN(CC([O-])=O)CC([O-])=O)=C1
Infrared spectrum
Conforms
Loss on drying
=<10.0 % (110°C, 1 h)
UV Lambda max
575 to 585 nm (0.1N NaOH)
Suitability
(for Zn titration) Passes test
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text