Silver chloride, 99+%, extra pure, Thermo Scientific Chemicals
Silver chloride, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver chloride, 99+%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 211270100
Price (USD)/ Each
73.65
Online exclusive
86.50 
Save 12.85 (15%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
73.65
Online exclusive
86.50 
Save 12.85 (15%)
Add to cart
Silver chloride, 99+%, extra pure, Thermo Scientific Chemicals
Catalog number211270100
Price (USD)/ Each
73.65
Online exclusive
86.50 
Save 12.85 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namesilver(1+) chloride
Molecular FormulaAgCl
InChI KeyHKZLPVFGJNLROG-UHFFFAOYSA-M
SMILES[Cl-].[Ag+]
Molecular Weight (g/mol)143.32
View more
SpecificationsSpecification SheetSpecification Sheet
Lead (Pb)=<10 ppm
Appearance (Color)White to pink-beige to grey
Iron (Fe)=<10 ppm
Appearance (Form)Powder
Copper (Cu)=<4 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only