Lithium nitrate, 99+%, extra pure, Thermo Scientific Chemicals
Lithium nitrate, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium nitrate, 99+%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
Plastic bottle
Catalog number 211320010
Price (USD)/ Each
218.65
Online exclusive
243.00 
Save 24.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
218.65
Online exclusive
243.00 
Save 24.35 (10%)
Add to cart
Lithium nitrate, 99+%, extra pure, Thermo Scientific Chemicals
Catalog number211320010
Price (USD)/ Each
218.65
Online exclusive
243.00 
Save 24.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7790-69-4
IUPAC Namelithium(1+) nitrate
Molecular FormulaLiNO3
InChI KeyIIPYXGDZVMZOAP-UHFFFAOYSA-N
SMILES[Li+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystalline powder
Titration after Ion exchange>=99.0 %
Appearance (Color)White
Loss on drying=<1 % (200°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only