Thermo Scientific Chemicals

Lithium nitrate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 211322500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Lithium nitrate, 99+%, extra pure, Thermo Scientific Chemicals

Catalog number: 211322500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 211322500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-69-4
IUPAC Name
lithium(1+) nitrate
Molecular Formula
LiNO3
InChI Key
IIPYXGDZVMZOAP-UHFFFAOYSA-N
SMILES
[Li+].[O-][N+]([O-])=O
Appearance (Form)
Adhering crystalline powder
Titration after Ion exchange
>=99.0 %
Appearance (Color)
White
Loss on drying
=<1 % (200°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text