Thermo Scientific™

Pierce™ High Sensitivity Streptavidin-HRP, Pre-Diluted

Catalog number: 21134
Thermo Scientific™

Pierce™ High Sensitivity Streptavidin-HRP, Pre-Diluted

Catalog number: 21134
Catalog Number
21134
Unit Size
2 mL
Price (EUR)
Availability
-
Quantity
Catalog NumberUnit SizePrice (EUR)AvailabilityQuantity
211342 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce High Sensitivity Streptavidin-HRP, 2 mL is an exclusive peroxidase-conjugated streptavidin biotin-binding protein that provides signal amplification like poly-HRP and exceptional storage stability.

Features of High Sensitivity Streptavidin-HRP, 2 mL:

High sensitivity—detect low levels of target without background; obtain high signal-to-noise ratios
Cost effective—use less conjugate in Western blotting and ELISA applications and still obtain excellent results
Flexible—compatible with typical chemiluminescent, fluorescent and colorimetric peroxidase substrates
Convenient—ready-to-use stabilized liquid format means there is no waiting for thawing and no need to aliquot

This specially-manufactured variety of HRP-conjugated streptavidin protein meets the demands of today's scientists for more sensitive detection in ELISA and Western blotting applications. The conjugate is suitable for use with chemiluminescent, chemifluorescent or colorimetric substrates. Each High Sensitivity HRP conjugate is packaged in an easy-to-use stabilized solution that enables convenient storage at 4°C for at least one year.

Our High Sensitivity HRP Conjugates outperform the competition on target detection, signal intensity and signal-to-noise ratios. Experiments indicate that the High Sensitivity Streptavidin-HRP can be diluted 8-to 10-fold more than traditional HRP conjugates and still produce similar results.

Related Products
Pierce™ High Sensitivity Streptavidin-HRP
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
2 mL
Target Molecule
Biotin
Specificity
Biotin
Detection Method
Colorimetric, Chemiluminescent, Chemifluorescent
Enzyme
HRP (Horseradish Peroxidase)
Form
Liquid
For Use With (Application)
Western Blotting, ELISA
Product Line
Pierce™
Label or Dye
HRP (Horseradish Peroxidase)

Contents & Storage

Storage at 4°C for at least one year.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text