Thermo Scientific Chemicals

Cadmium iodide, 99%, Thermo Scientific Chemicals

Catalog number: 211462500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Cadmium iodide, 99%, Thermo Scientific Chemicals

Catalog number: 211462500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 211462500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7790-80-9
IUPAC Name
cadmium(2+) diiodide
Molecular Formula
CdI2
InChI Key
OKIIEJOIXGHUKX-UHFFFAOYSA-L
SMILES
[Cd++].[I-].[I-]
Appearance (Color)
White to light yellow
Titration Complexometric
>=98.5 %
Appearance (Form)
Powder or crystals and/or flakes

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text